Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
NEBNext® rRNA Depletion Kit (Bacteria), New England Biolabs
Supplier: New England Biolabs
NEBNext®
E7850X E7850L E7850S E7850S E7850X E7850L
76407-336EA 4042 USD InStock
76407-336 CA76407-334 CA76407-336 76407-332 76407-334 CA76407-332
NEBNext® rRNA Depletion Kit (Bacteria), New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext rRNA Depletion Kit (Bacteria) employs the NEBNext RNase H-based RNA depletion workflow to target removal of rRNA from gram-positive and gram-negative organisms. The method is effective with intact and degraded RNA, from monocultures or samples with mixed bacterial species (e.g., metatranscriptomic).
  • Efficient, specific depletion of bacterial rRNA (5S, 16S, 23S)
  • Compatible with both gram-positive and gram-negative organisms
  • Effective with monocultures and mix of bacterial species (e.g., metatranscriptome)
  • Compatible with a broad range of input amounts: 10 ng - 1 µg
  • Suitable for low-quality or high-quality RNA
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty