Would you like to log in again?
NEBNext® rRNA Depletion Kit (Bacteria) with RNA Sample Purification Beads, New England Biolabs
Supplier: New England Biolabs
NEBNext®
E7860L E7860X E7860L E7860X E7860S E7860S
76407-340EA 317 USD InStock
76407-340 CA76407-338 76407-338 76407-342 CA76407-340 CA76407-342
NEBNext® rRNA Depletion Kit (Bacteria) with RNA Sample Purification Beads, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext rRNA Depletion Kit (Bacteria) employs the NEBNext RNase H-based RNA depletion workflow to target removal of rRNA from gram-positive and gram-negative organisms. The method is effective with intact and degraded RNA, from monocultures or samples with mixed bacterial species (e.g., metatranscriptomic). For convenience, this kit includes RNA Clean beads.
  • Efficient, specific depletion of bacterial rRNA (5S, 16S, 23S)
  • Compatible with both gram-positive and gram-negative organisms
  • Effective with monocultures and mix of bacterial species (e.g., metatranscriptome)
  • Compatible with a broad range of input amounts: 10 ng - 1 µg
  • Includes Agencourt RNAClean XP beads
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty