Would you like to log in again?
Hi-T7 RNA Polymerase (High Concentration), New England Biolabs
Supplier: New England Biolabs
M0470T M0470T
76407-350EA 714 USD InStock
76407-350 CA76407-350
Hi-T7 RNA Polymerase (High Concentration), New England Biolabs
Nucleic Acid Reagents RNA Synthesis and Transcription
Hi-T7 RNA Polymerase (High Concentration) is offered at a 20-fold higher concentration than our standard Hi-T7 RNA Polymerase and is ideal for experienced users interested in building and optimizing their own in vitro transcription reactions.
  • Active from 37 to 56 °C, optimal incubation temperature is 50 to 52 °C
  • In vitro transcription using T7 phage promoter
  • High concentration version of Hi-T7 RNA Polymerase
The enzyme is accompanied by Hi-T7 RNA Polymerase reaction buffer and Magnesium chloride solution to further enable testing of different conditions such as nucleotide concentrations. In vitro transcription involves multiple components in addition to the RNA Polymerase including ribonucleotides, inorganic pyrophosphatase and RNase inhibitor, which can be purchased separately. Please refer to the Related Products section for details. Hi-T7 RNA Polymerase is an engineered DNA-dependent RNA polymerase that is highly specific for T7 phage promoters and is designed to function at higher temperatures than the wild-type bacteriophage T7 RNA Polymerase.
Concentration = 1000000 units/ml.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty