Would you like to log in again?
Hi-T4 DNA Ligase
Supplier: New England Biolabs
M2622S M2622L M2622L M2622S
76407-352EA 291 USD InStock
76407-352 CA76407-352 76407-354 CA76407-354
Hi-T4 DNA Ligase
Enzymes
A thermotolerant variant of T4 DNA Ligase, Hi-T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini in duplex DNA or RNA and is designed to function at higher temperatures than wild type T4 DNA Ligase.
  • Hi-T4 DNA Ligase is an engineered variant of T4 DNA Ligase with improved thermostability
  • Capable of joining blunt end and cohesive end termini
  • Capable of repairing single stranded nicks in duplex DNA and some DNA/RNA hybrids
  • Supplied with both T4 DNA Ligase buffer and StickTogether DNA Ligase buffer
Hi-T4 DNA Ligase will join blunt end and cohesive end termini as well as repair single-stranded nicks in duplex DNA, RNA or DNA/RNA hybrids at temperatures as high as 50 °C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty