Would you like to log in again?
Luna® Cell Ready Probe One-Step RT-qPCR Kit, New England Biolabs
Supplier: New England Biolabs
Luna®
E3031S E3031S
76407-358EA 1061 USD InStock
76407-358 CA76407-358
Luna® Cell Ready Probe One-Step RT-qPCR Kit, New England Biolabs
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
The Luna® Cell Ready Probe One-Step RT-qPCR Kit provides all the necessary components for direct, probe-based RNA detection and quantitation, bypassing the need for RNA extraction and purification.
  • Contains reagents for 100 lysis preps and 500 one-step RT-qPCR reactions (probe)
  • Effective cell lysis preparation from 10 to 100000 cells across numerous cell lines
  • Coordinated cell lysis, RNA release, and genomic DNA removal in a fast 15-minute protocol
  • Luna WarmStart RT paired with Hot Start Taq for increased thermostability and room temperature setup
Coordinating the actions of DNase I and the Luna Cell Ready Protease, the Luna Cell Ready Lysis Module offers a simple alternative workflow resulting in effective cell lysis, RNA release, and genomic DNA removal simultaneously in a 15-minute protocol. The Lysis Module includes a unique Luna Cell Ready RNA Protection Reagent that maintains RNA integrity during cell lysis. The lysis capacity spans 10 to 100000 cells in a 50 µl lysis reaction. Up to 2 µl of lysate (equivalent to RNA from 0.2 to 4000 cells) can be transferred into 20 µl downstream RT-qPCR reactions.
Delivery information: The Luna Cell Ready Probe One-Step RT-qPCR Kit is composed of:
1) The Luna Cell Ready Lysis Module
2) The Luna Universal Probe One-Step RT-qPCR Kit.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty