Would you like to log in again?
Luna Cell Ready Lysis Module, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
E3032S E3032S
76407-360EA 571 USD InStock
76407-360 CA76407-360
Luna Cell Ready Lysis Module, New England Biolabs
Buffers Extraction Buffers
Cell cultures are often analyzed for gene expression or treatment responses as a proxy for a living organism. Traditionally, RNA is extracted and purified from treated cells via column-based or chemical methods. The Luna Cell Ready Lysis Module is a fast, convenient, and sensitive alternative for evaluating RNA expression levels without performing traditional RNA extraction steps.
  • Coordinated cell lysis, RNA release, and genomic DNA removal in a fast 15-minute protocol
  • Effective cell lysis preparation from 10 to 100000 cells across numerous cell lines
  • Minimal sample loss compared to alternative RNA extraction methods, especially at low cell input
  • Compatible with high-throughput applications: cell lysis can take place at room temperature
The module coordinates cell lysis, RNA release, and genomic DNA removal simultaneously, resulting in cell lysate ready for direct RT-qPCR analysis with Luna Universal One-Step RT-qPCR kits. Similar to other Luna products, the lysis buffer includes an inert blue tracking dye for visual assistance throughout the workflow.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty