Would you like to log in again?
NEBExpress Cell-Free E. coli Protein Synthesis System, New England Biolabs
Supplier: New England Biolabs
NEBExpress
E5360S E5360S E5360L E5360L
76407-362EA 1922 USD InStock
76407-362 76407-364 CA76407-362 CA76407-364
NEBExpress Cell-Free E. coli Protein Synthesis System, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The NEBExpress Cell-free E. coli Protein Synthesis System is an extract-based transcription/translation system derived from E. coli cells that have been engineered for high in vitro synthesis performance. It is designed to synthesize proteins ranging in size from 17 to 230 kDa in just 2 to 4 hours under the control of T7 RNA Polymerase.
  • Synthesize high yields of protein (typically 0.5 mg/ml)
  • Protein can be synthesized and visualized in approximately 2 to 4 hours
  • Templates can be plasmid DNA, linear DNA or mRNA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty