Would you like to log in again?
TUBING APSPG SILICONE 0.063 X 0.188 IN
Supplier: NEWAGE INDUSTRIES. INC
Tubing, APSPG platinum-cured silicone, specially formulated to provide outstanding compression characteristics,excellent flex fatigue,tear resist, extended life in peristaltic pump applications, low volatile grade, Dimensions: 0.063 X 0.188 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBING APSPG SILICONE 0.063 X 0.188 IN 76407-450 A7020130-25 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty