Would you like to log in again?
TUBING APST SILICONE 0.030 X 0.160 IN
Supplier: NEWAGE INDUSTRIES. INC
Tubing, APST, translucent, platinum-cured USP Class VI silicone,extruded in a Class 1000 clean room & stored in a particulate-controlled warehouse, highly pure product meets FDA,EP,3A,ISO standards, Dimensions: 0.030 X 0.160 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBING APST SILICONE 0.030 X 0.160 IN 76407-454 A7000591-25 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty