Would you like to log in again?
GLOVES PVC/NITRILE BLUE SZ 9 12IN PK12
Supplier: Ansell Healthcare
Gloves Pvc/Nitrile Blue Sz 9 12In PK12^ AlphaTec* Gloves, Color: Blue, Material: PVC/Nitrile, Guantlet Cuff, Silicone Free, Chemcial/Liquid Splash Resistant Glove/Food Handling/Cold Food Storage, Size: 9, Lenght: 12 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES PVC/NITRILE BLUE SZ 9 12IN PK12 76407-482 04002090 Case of 72 (1/ Itm/Bx) Retrieving
Pack of 12 (1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty