Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Packing Strips, DUKAL™ Corporation
Supplier: Dukal
271 270 273 272 263 260 261
76407-538CS 62.3 USD InStock
76407-538 76407-544 76407-536 76407-540 76407-530 76407-542 76407-532
Packing Strips, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Sterile packing strips are deisgned for use in wound care management applications such as nasal or sinus packing.
  • Made of a woven gauze fabric which reduces linting and fraying
  • Strips are packed in unbreakable poly bottles with tamper evident seals
  • No natural rubber latex
Packing strips are available with or without an Iodoform treatment, which provides odor-resistance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty