Would you like to log in again?
Conforming Stretch Gauze, DUKAL™ Corporation
Supplier: Dukal
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
704 703 603 601 702 701 606 706 602 604
76407-580BG 6.72 USD InStock
76407-580 76407-592 76407-582 76407-594 76407-590 76407-578 76407-596 76407-574 76407-598 76407-576
Conforming Stretch Gauze, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Rayon/poly knitted stretch gauze bandage.
  • Specifically designed not to slip while in use
  • Available in non sterile and sterile formats
  • No natural rubber latex
Conforming stretch gauze provides slight compression to the site while securely holding to any body contour. The unique weave construction and finished edges prevent unraveling and linting.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty