Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Transparent Semi-Permeable Dressings, DUKAL™ Corporation
Supplier: Dukal
4112 4045 4152
76407-622BX 73.91 USD InStock
76407-622 76407-650 76407-652
Transparent Semi-Permeable Dressings, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Micro porous transparent film with a high Moisture Vapor Transmission Rate (MVTR).
  • Ideal for IV sites, minor burns, ulcers, skin tears, and donor sites
  • Easy two-step application
  • Rounded edges prevent roll up
  • Removable patient label for chart
  • No natural rubber latex
Dressings protect against fluid contamination and bacteria on and around the wound site.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty