Would you like to log in again?
Sterile Post-Op Sponges, DUKAL™ Corporation
Supplier: Dukal
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5444 5434
76407-666BX 7.47 USD InStock
76407-666 76407-664
Sterile Post-Op Sponges, DUKAL™ Corporation
Wipes Sponges
A multi-layer, woven gauze-faced sponge with cellulose fill.
  • Well suited for use in place of a single traditional sponge
  • No natural rubber latex
Post-Op sponges provide fast wicking and increased absorbency.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty