Would you like to log in again?
Type VII X-Ray Detectable Gauze, DUKAL™ Corporation
Supplier: Dukal
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4824-2X 416-2X 432-2X 488-2X 4412-2X 4432-2X 4416-2X 482-2X 4812-2X
76407-926CS 329.2 USD InStock
76407-926 76407-928 76407-907 76407-922 76407-934 76407-924 76407-930 76407-932 76407-910
Type VII X-Ray Detectable Gauze, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Blue filament runs through each sponge and has a unique wave pattern to ensure easy X-ray detection.
  • USP Type VII gauze, 100% woven-cotton
  • Sterile sponges are packaged in rigid plastic tray with peel back lid
  • No natural rubber latex
Both sterile and non-sterile sponges are banded in 10’s to ensure accurate count.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty