Would you like to log in again?
BZK Towelettes, DUKAL™ Corporation
Supplier: Dukal
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
855-1000 855-1200 855
76407-966CS 87.36 USD InStock
76407-966 76407-964 76289-522
BZK Towelettes, DUKAL™ Corporation
Wipes Disposable Wipes and Towels Disinfectant Wipes
BZK Towelettes allow for antiseptic cleansing of face, hands, and body without the need for soap and water.
  • Individually sealed foil packs keep towelettes moist
  • Alcohol-free
  • No natural rubber latex
Common applications include cleaning of skin around minor cuts, scrapes, and burns, as well as cleansing prior to clean catch urine tests.
Packaging: Available in different case sizes.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty