Would you like to log in again?
SELF-STICKING SAFETY XRF SEC FILM PK25
Supplier: CHEMPLEX INDUSTRIES
Self-Sticking Safety XRF Secondary Film^ Spectrofilm* Self-Sticking Safety XRF Secondary Film, to protect the x-ray tube and detector windows and related electronics from exposure to accidental sample material spillage.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SELF-STICKING SAFETY XRF SEC FILM PK25 76408-052 2042R35P4.0 Pack of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty