Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AlphaTec® CSM Isolator Gloves, 85-304 Series, Ansell

Supplier: Ansell

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AlphaTec®
85304110 85304095
76408-476CS 6731.54 USD InStock
76408-476 76408-478
AlphaTec® CSM Isolator Gloves, 85-304 Series, Ansell
Gloves
Non-clean/non sterile CSM RABS/Isolator gloves for use in 305 mm (12") ports.

  • Designed for high resistance against concentrated acids and bases
  • Port size: 305 mm (12")
  • Soft, flexible CSM material designed for ease of use
  • Resists ageing from exposure to oxygen, UV rays, and ozone
  • White colouring for easy detection of contamination

AlphaTec® CSM RABS and Isolator gloves are designed for high resistance against concentrated acids and bases. They are made from soft, flexible material designed for ease of use.

Certifications: CE 0493, EN 374:2003, EN ISO 374-5:2016, EN 388:2003, EN 388:2016 +A1:2018, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, Category III, UKCA 0321.

Packaging: 1 pair per black sealed plastic bag, 10 pairs per shipper carton.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty