Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AlphaTec® EPDM Isolator Gloves, 85-502 Series, Ansell

Supplier: Ansell
AlphaTec®
85502095 85502110
76408-494CS 9471.6 USD InStock
76408-494 76408-492
AlphaTec® EPDM Isolator Gloves, 85-502 Series, Ansell
Gloves
Non-clean/non sterile EPDM RABS/Isolator gloves for use in 254 mm (10") ports.

  • Designed for repeated autoclave sterilization (up to 50 times)
  • Port size: 254 mm (10")
  • Comfortable and dexterous with great tactile sensitivity
  • Resistant against hydrogen peroxide solutions and common disinfecting chemicals
  • Resists aging from exposure to oxygen, UV rays, and ozone
  • Halogen-free: suitable for disposal by incineration

AlphaTec® EPDM RABS and Isolator gloves are designed from high quality material, in compliance with FDA CFR21. Comfortable and dexterous, with great tactile sensitivity. Designed for repeated autoclave sterilization, reducing the need for glove replacements.

Certifications: CE 0493, EN 374:2003, EN 388:2003, EN 388:2016, EN 388:2016 +A1:2018, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016, EN 420:2003 + A1:2009, Category III.

Packaging: 1 pair per black sealed bag, 10 pairs in black sealed bags per shipper case.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty