Would you like to log in again?
VWR® Precision Grip Gloves, NBR Coating
Supplier: VWR
VWR®
GCVWRWRKA1NB10 GCVWRWRKA1NB09 GCVWRWRKA1NB06 GCVWRWRKA1NB11 GCVWRWRKA1NB08 GCVWRWRKA1NB07
76408-582CS 706.06 USD InStock
76408-582 76408-590 76408-580 76408-586 76408-584 76408-588
VWR® Precision Grip Gloves, NBR Coating
Gloves
The VWR® precision grip glove with nitrile coating provide basic cut protection and grip. Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment.
  • Nylon composition
  • Nitrile (NBR) coated
  • Blue knit glove with black coating
Also great for package handling, picking, and packing.
Certifications: Meets requirements for ANSI A1 Cut Protection (Level A). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty