Would you like to log in again?
Vivacell® 100 Ultrafiltration Units, Sartorius
Supplier: Sartorius
Vivacell 100
VC1072 VC1062 VC1012 VC1052 VC1031 VC1071 VC1041 VC1051 VC1061 VC1031 VC1022 VC1001 VC1002 VC1021 VC1042 VC1011 VC1032
76408-626PK 471.74 USD InStock
76408-626 76408-616 76408-624 76408-614 76408-608 CA76408-618 76408-628 76408-606 76408-618 76408-630 76408-622 76408-634 76408-612 76408-620 76408-632 76408-610 76409-428
Vivacell® 100 Ultrafiltration Units, Sartorius
Filters Centrifugal Filters
Vivacell® 100 is a disposable ultrafiltration unit for the concentration of samples up to 98 ml that can be used either in a centrifuge or for pressure filtration.
  • Easy to handle
  • High flux PES membrane
  • Vertical twin membrane for ultra-fast concentration
  • High retentate recovery >90%
  • Total process flexibility
  • Compatible with swing out rotors
Vivacell® 100 combines the ease of use of centrifugal devices with the flexibility and control provided by pressurized ultrafiltration cells. Vivacell® 100 is easy to assemble, requires no tubing connections or stirring mechanisms and can be adapted to equipment availability or to specific user preferences.
For convenience, the units are simply spun in a large capacity centrifuge (carriers accepting 250 ml bottles). For better process control, gas pressure can be combined with gentle orbital shaking. For highest simplicity of use with minimum equipment requirements, the device is pressurized and then left standing on a bench top or in a refrigerator. The concentrator features a vertical membrane that minimizes fouling, while the built-in deadstop of 350 µl will prevent concentration to dryness.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty