Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Vivaflow® 50R Ultrafiltration Units, Sartorius
Supplier: Sartorius
Vivaflow®
VF05H4 VF05H0 VF05H1 VF05H4 VF05H0 VF05H2 VF05H1 VF05H2
76408-658EA 340.83 USD InStock
CA76408-658 CA76408-656 76408-658 CA76408-660 76408-660 CA76408-654 76408-656 76408-654
Vivaflow® 50R Ultrafiltration Units, Sartorius
Filters Centrifugal Filters
These are for 0.1 to 1000 ml, Hydrosart® membrane. Vivaflow 50R is a ready to use and reusable laboratory crossflow cassette for applications like cell culture supernatant concentration, virus concentration and water sample concentration. The thin channel, flip flow recirculation path of the modules provides high crossflow velocities with minimum pump speed requirements. A single module with 50 cm² active membrane area typically concentrates 250 ml by a factor of 20 in less than 30 min.
  • Fast and easy protein sample concentration
  • Reusable and optimal for cell culture supernatant and virus concentration
  • Plug and play laboratory cross flow cassette
Materials of construction are acrylic main housing and flow channel, PP membrane support, silicone seal and O-rings, pressure indicator (PP) SS** spring, PP flow restrictor, nylon fittings, PVC (medical grade) tubings.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty