Would you like to log in again?
VIVAFLOW 50 1000000 MWCO PES PK2
Supplier: SARTORIUS CORPORATION EDI
Vivaflow* 50 cross flow cassette, Molecular weight cutoff: 1000000, Material: PES, Plug and play laboratory cross flow cassette, Modular - scalable to 6 units, Disposable, High flow rate and recovery, Optimal for aqueous sample concentration e.g, protein and nanoparticle, size: 100ml-3l
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIVAFLOW 50 1000000 MWCO PES PK2 76408-672 VF05P6 Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty