Would you like to log in again?
Vivaflow® 200 Modular Tangential Flow System, Sartorius
Supplier: Sartorius
Vivaflow®
VFS204 VFS204
76408-736EA 2950.45 USD InStock
76408-736 CA76408-736
Vivaflow® 200 Modular Tangential Flow System, Sartorius
Filters Tangential Flow Systems
Vivaflow® 200 is a ready to use and reusable laboratory crossflow cassette for concentrating up to 5 L samples.
  • Optimal for cell culture supernatant and virus concentration
  • High flow rate and recovery
The membrane surface per device is 200 cm². One unit comes with all necessary accessories for running the device with a laboratory pump and a size 16 pump head.
For speeding up the concentration, two cassettes can be run simultaneously.
Packaging: Pump, easy load pump head (size 16), tubing, 500 ml sample/diafiltration reservoir.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty