Would you like to log in again?
Vivacon® 500, Centrifugal Concentrators, Sartorius
Supplier: Sartorius
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Vivacon®
VN01H32 VN01H31 VN01H91 VN01H21ETO VN01H01 VN01H81 VN01HL12 VN01H42 VN01H92 VN01H91 VN01HL12 VN01H02 VN01HH12 VN01H32 VN01H42 VN01H82 VN01H41 VN01H41 VN01H02 VN01H01 VN01H92 VN01H22 VN01H22 VN01H21 VN01H21 VN01H31 VN01H82 VN01H81 VN01H41ETO
76408-772PK 574.9 USD InStock
CA76408-768 CA76408-746 CA76408-788 CA76408-744 76408-772 76408-750 CA76408-760 CA76408-782 76408-770 CA76408-780 76408-776 CA76408-764 CA76408-786 CA76408-742 CA76408-784 76408-780 76408-748 CA76408-776 76408-768 76408-746 76408-784 76408-782 76408-760 76408-788 76408-744 CA76408-772 CA76408-750 76408-742 76408-764
Vivacon® 500, Centrifugal Concentrators, Sartorius
Filters Centrifugal Filters
Vivacon® 500 centrifugal concentrators have been designed for DNA and protein concentration and rebuffering. They feature a horizontal membrane design that is preferred by some users for reasons of consistency with previously used procedures and reproducibility.
  • Optimal performance with very dilute samples, e.g. forensic specimens
  • Low binding materials; high recoveries even from diluted samples
  • Re-spin possibility; complete and highly reproducible sample recovery
  • Volume graduation and the molecular weight cut-off printed on individual devices
  • For samples up to 500 µl membrane: Regenerated cellulose Hydrosart®
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty