Would you like to log in again?
Vivacon® 2 Centrifugal Concentrators, Sartorius
Supplier: Sartorius
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Vivacon®
VN02H92 VN02H92 VN02H02 VN02H33 VN02H23 VN02H22 VN02H01 VN02H43 VN02H01 VN02H41 VN02H42 VN02H02 VN02H42 VN02H41 VN02H03 VN02H43 VN02H31 VN02H22 VN02H93 VN02H32 VN02H91 VN02H21 VN02H32 VN02H21 VN02H23 VN02H31 VN02H91
76408-836PK 737.11 USD InStock
CA76408-800 CA76408-822 76408-836 76408-812 76408-834 CA76408-826 CA76408-804 76408-818 76408-792 76408-796 CA76409-306 CA76408-808 CA76408-834 CA76408-812 76408-822 76408-800 CA76408-838 76408-804 76408-826 CA76408-836 CA76408-792 CA76408-790 CA76408-796 76408-790 76408-808 CA76408-818 CA76409-310
Vivacon® 2 Centrifugal Concentrators, Sartorius
Filters Centrifugal Filters
Vivacon® 2 centrifugal concentrators have been designed for DNA concentration, buffer exchange applications and primer removal.
  • Optimal performance with very dilute samples for example, forensic specimens
  • Low binding materials
  • High recoveries even from diluted samples
  • Re-spin possibility
  • Complete and highly reproducible sample recovery
  • Volume graduation and the molecular weight cut-off printed on individual devices
For samples up to 2 ml with regenerated cellulose Hydrosart® membrane. They feature a horizontal membrane design that is preferred by some users for reasons of consistency with previously used procedures and reproducibility.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty