Would you like to log in again?
Vivaspin® 500 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Vivaspin®
VS01S1 VS0132 VS0191 VS0141 VS0111 VS0151 VS0102 VS0121 VS0111 VS01S1 VS0191 VS0172 VS0132 VS0192 VS0101 VS0161 VS0142 VS0121 VS0141 VS0151 VS0131 VS0122 VS0171 VS0142 VS0162 VS0122 VS0152 VS0162 VS0112 VS0192 VS0171 VS0131 VS0102 VS0101 VS0172 VS0161 VS0112 VS0152
76408-878PK 163.34 USD InStock
CA76408-866 CA76408-888 CA76408-886 76408-878 CA76408-868 CA76408-880 76408-894 76408-872 76408-870 76408-892 CA76408-884 76408-898 76408-876 CA76408-882 76408-874 76408-896 76409-430 76409-432 76408-868 CA76408-878 76408-888 76408-866 CA76408-898 CA76408-876 76408-882 CA76408-870 CA76408-892 76408-880 CA76408-890 76408-886 CA76408-896 CA76408-874 76408-884 CA76408-894 CA76408-872 76408-890 CA76409-430 CA76409-432
Vivaspin® 500 Centrifugal Filtration Units, Sartorius
Filters Centrifugal Filters
Vivaspin® centrifugal concentrators are disposable ultrafiltration devices for the concentration of biological samples. Vivaspin® 500 centrifugal filter units offer a simple, one step procedure for sample concentration with volumes from 5 to 500 µl. They can effectively be used in fixed angle rotors accepting 2.2 ml centrifuge tubes.
  • High retentate recovery >90%
  • Vertical membrane design for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
The vertical membrane design and thin channel filtration chamber (US 5,647,990), minimises membrane fouling and provides high speed concentrations.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty