Would you like to log in again?
Vivaspin® 2 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Vivaspin®
VS02H92 VS0261 VS0291 VS0272 VS0222 VS02H02 VS0252 VS0222 VS02V1 VS02X1 VS02H02 VS0252 VS0291 VS0251 VS0212 VS02H21 VS02H01 VS0201 VS02H22 VS02S1 VS0202 VS02V1 VS02U1 VS0251 VS0242 VS0231 VS0241 VS0221 VS02H12 VS02X1 VS0272 VS02H21 VS0231 VS02V1 VS0271 VS0211 VS0202 VS02X2 VS0262 VS0241 VS02X2 VS02V2 VS02H91 VS02H11 VS0271 VS02H92 VS02V2 VS02H11 VS02H91 VS0261 VS0212 VS0232 VS02H22 VS02H12 VS0262 VS02H01 VS0232 VS0221 VS0211 VS0292 VS02U2 VS0201 VS0292 VS0242
76408-934PK 636.99 USD InStock
76408-934 CA76408-944 CA76408-900 76408-932 CA76408-942 76408-938 CA76408-904 CA76408-948 76408-936 CA76408-946 CA76408-902 76408-930 CA76408-940 CA76408-908 CA76408-906 CA76408-910 76408-902 76408-946 CA76408-952 76408-944 76408-900 CA76408-958 CA76408-914 CA76408-959 76408-906 CA76408-912 76408-948 76408-904 CA76408-950 76408-942 76408-940 CA76408-918 CA76408-916 76408-908 76408-912 76408-956 CA76408-922 76408-910 76408-954 CA76408-920 CA76408-964 76408-916 CA76408-926 76408-958 76408-914 CA76408-924 CA76408-962 76408-950 CA76408-960 76408-918 CA76408-928 CA76408-932 76408-924 CA76408-930 76408-922 CA76408-936 76408-928 CA76408-934 76408-926 76408-960 76408-920 76408-964 76408-962 CA76408-938
Vivaspin® 2 Centrifugal Filtration Units, Sartorius
Filters Centrifugal Filters
Vivaspin® concentrators are disposable ultrafiltration devices for the concentration of biological samples of 0.4 to 2 ml.
  • High retentate recovery >90%
  • Vertical membrane design for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
Vivaspin® 2 can effectively be used in swing bucket or fixed angle rotors accepting 15 ml centrifuge tubes. The patented vertical membrane design and thin channel filtration chamber (US 5647990) minimizes membrane fouling and provides high speed concentrations, even with particle-laden solutions.
Vivaspin® 2 combines the speed of the classic Vivaspin® products with low internal surface and membrane area in order to achieve superior recoveries from very dilute solutions.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty