Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Vivaspin® Turbo 4 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
Vivaspin®
VS04T01 VS04T32 VS04T12 VS04T91 VS04T02 VS04T42 VS04T22 VS04T31 VS04T01 VS04T32 VS04T92 VS04T21 VS04T41 VS04T12 VS04T02 VS04T91 VS04T11 VS04T42 VS04T11 VS04T31 VS04T41 VS04T21 VS04T22 VS04T92
76408-978PK 212.32 USD InStock
76408-978 CA76408-976 76408-968 CA76408-966 CA76408-988 76408-976 CA76408-974 76408-966 CA76408-986 76408-988 CA76408-978 CA76408-968 76408-970 CA76408-980 76408-982 76408-980 76408-974 CA76408-972 76408-986 CA76408-984 76408-972 CA76408-970 76408-984 CA76408-982
Vivaspin® Turbo 4 Centrifugal Filtration Units, Sartorius
Filters Centrifugal Filters
Vivaspin® Turbo 4 can handle 0.04 to 4 ml sample volume in swing-out and fixed angle rotors accepting 15 ml centrifuge tubes.
  • High retentate recovery >95%
  • pH 1 to 14 suited PS housing with high chemical compatibility
  • Safe sample concentration and easy recovery due to unique, angular and pipette friendly dead-stop pocket
  • Volume graduation for easy detection/monitoring of the concentrate volume
The Vivaspin® Turbo 4 optimised design, the sleek internal profile, ensures maximum process speeds right the way down to the last few micro litres after >100 fold concentration.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty