Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Vivaspin® 6 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
Vivaspin®
VS0622 VS0621 VS0691 VS0691 VS0661 VS0602 VS0642 VS0631 VS0671 VS0641 VS0652 VS0652 VS0692 VS0671 VS0601 VS0692 VS0602 VS0612 VS0611 VS0641 VS0601 VS0651 VS0612 VS0631 VS0662 VS0672 VS0651 VS0622 VS0661 VS0672 VS0632 VS0632 VS06S1 VS0642 VS0611 VS0662 VS0621
76408-998PK 206.89 USD InStock
76408-998 76409-020 76408-992 76408-990 76408-996 76408-994 76409-018 CA76409-006 CA76409-008 76409-010 CA76409-020 76409-012 CA76409-000 CA76409-022 76409-014 CA76409-024 CA76409-002 76409-016 CA76409-004 CA76409-026 CA76408-998 CA76408-996 CA76408-990 CA76408-994 CA76408-992 CA76409-016 76409-008 CA76409-018 76409-022 76409-000 CA76409-010 76409-002 76409-024 CA76409-012 76409-004 CA76409-014 76409-006
Vivaspin® 6 Centrifugal Filtration Units, Sartorius
Filters Centrifugal Filters
Vivaspin® 6 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin® 6 can process 6 ml in either swing bucket or fixed angle rotors accepting standard 15 ml conical bottom test tubes.
  • High retentate recovery <90%
  • Vertical membrane design for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
Vivaspin® 6 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. The remaining volume is easy to read off the printed scale on the side of the concentrator and the modified dead stop pocket further simplifies direct pipette recovery of the final concentrate.
Vivaspin® 6 is optimally suited for the concentration of antibody-, protein-, nanoparticle- and virus samples from 6 ml sample volume to 30 µl in a centrifuge accepting 50 ml conical bottom centrifuge tubes. These are the only device of its size that can operate 6 ml sample.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty