Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sartorius Vivaspin® Turbo 15

Supplier: Sartorius

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Vivaspin®
VS15T01 VS15T92 VS15T92 VS15T21 VS15T91 VS15T32 VS15T42 VS15T22 VS15T02 VS15T91 VS15T32 VS15T31 VS15T11 VS15T22 VS15T42 VS15T21 VS15T41 VS15T11 VS15T02 VS15T31 VS15T41 VS15T01 VS15T12 VS15T12
76409-060PK 791.58 USD InStock
76409-060 76409-050 76409-062 CA76409-060 76409-052 CA76409-050 76409-042 76409-064 CA76409-062 CA76409-052 76409-054 CA76409-064 76409-044 CA76409-042 76409-056 CA76409-054 76409-046 CA76409-044 CA76409-056 76409-058 CA76409-046 76409-048 CA76409-058 CA76409-048
Sartorius Vivaspin® Turbo 15
Filters Centrifugal Filters
The Vivaspin® Turbo 15 can handle up to 15 ml sample volume in swing bucket rotors and 11 ml in fixed angle rotors accepting 50 ml centrifuge tubes.

  • High chemical resistance
  • High retentate recovery: more than 95%
  • Dead stop volume: 60
  • High flux PES membrane
  • Universal rotor compatibility

The UV joining technology creates a smooth joint transition between membrane and plastic housing, which allows the collection of the complete concentrated sample into the pipette-friendly dead stop pocket.

The unit features a polystyrene butadiene copolymer body and a polyethersulfone membrane with a polystyrene cap.

The Vivaspin® Turbo 15’s sleek internal profile ensures maximum process speeds down to the last few microliters after 100-fold concentration.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty