Would you like to log in again?
Vivaspin® 20 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Vivaspin®
VS2061 VS2042 VS2052 VS2041 VS2031 VS2022 VS2001 VS2072 VS2012 VS2032 VS2031 VS2051 VS2002 VS2021 VS2011 VS20S1 VS2092 VS2061 VS2041 VS2071 VS2022 VS2032 VS2011 VS2072 VS2051 VS2071 VS2092 VS2062 VS2021 VS2042 VS2001 VS2002 VS2012 VS2091 VS2091 VS2062 VS2052
76409-080PK 743.72 USD InStock
76409-080 CA76409-090 76409-082 CA76409-070 CA76409-092 76409-084 CA76409-072 CA76409-094 76409-086 76409-076 76409-098 CA76409-086 76409-078 CA76409-088 CA76409-066 CA76409-100 CA76409-068 76409-090 76409-070 76409-092 CA76409-080 76409-094 76409-072 CA76409-082 76409-096 76409-074 CA76409-084 CA76409-096 CA76409-074 76409-066 76409-088 CA76409-076 76409-100 CA76409-098 76409-068 CA76409-078 76409-102
Vivaspin® 20 Centrifugal Filtration Units, Sartorius
Filters Centrifugal Filters
Vivaspin® 20 centrifugal concentrators offer increased volume flexibility and performance to the Vivaspin® potfolio. Vivaspin® 20 handles 5 to 20 ml in swing bucket rotors and 14 ml in fixed angle rotors accepting 50 ml centrifuge tubes. Vivaspin® 20 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. Volume graduation allows monitoring the concentration process and the modified dead stop pocket simplifies pipetting of the final concentrate.
  • High retentate recovery >90%
  • Vertical twin membrane for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
  • Polycarbonate body and filterate vessel, polypropylene concentrator cap with acetal or aluminum pressure head
For higher process flexibility, it is available with unique accessories and operating methods. For one-step desalting following concentration, a diafiltration cup filled with buffer can be placed on the concentrator and spun another time to achieve 98% salt removal, instead of two spins to achieve the same result with the traditional refill and re-spin procedure.
When an appropriate centrifuge is unavailable, or for single sample processing, for gas pressure filtration, it can be filled with up to 15 ml and then pressurised for bench top concentration. For even faster processing, gas pressure filtration can be combined with centrifugation. 'Pressure-fugation' is particularly useful for difficult or viscous samples such as serum, when using a low process temperature, or generally when minimum process time is essential.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty