Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Vivapure® Ion Exchange (IEX) Mini/Maxi, Protein Purification Spin Columns, Sartorius

Supplier: Sartorius
Vivapure®
VS-IX01SH24 VS-IX01QH24 VS-IX20SH08 VS-IX01SH24 VS-IX20SH08 VS-IX20QH08 VS-IX20DH08 VS-IX01DH24 VS-IX01SQ16 VS-IX01DH24 VS-IX20QH08 VS-IX20DH08 VS-IX01SQ16 VS-IX01QH24
76409-130PK 510.83 USD InStock
76409-130 76409-128 CA76409-126 CA76409-128 76409-120 76409-132 CA76409-130 CA76409-120 76409-122 CA76409-132 CA76409-122 76409-124 CA76409-124 76409-126
Vivapure® Ion Exchange (IEX) Mini/Maxi, Protein Purification Spin Columns, Sartorius
Filters Centrifugal Filters
Vivapure® Ion Exchange (IEX) spin colums are centrifugal devices. They incorporate Sartobind membrane adsorber technology as their chromatography matrix. Sartobind IEX membrane adsorbers are based on stabilised regenerated cellulose and display a microporous structure with a pore size of >3 μm, which is orders of magnitude larger than conventional chromatographic gel materials.

  • Ready to use
  • Replaces time-consuming column chromatography protein purification
  • Scalable to industrial scale applications
  • Available in Mini and Maxi formats
  • Ideal for sample preparation before 1D or 2D PAGE, pre-fractionation and purification of various proteins

The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many different samples are processed in parallel.

Mini format: for binding up to 4 mg (500 µl capacity)
Maxi format: for binding up to 60 to 80 mg (20 ml capacity)

Strong acidic cation exchanger: Sulphonic acid (S)
Strong basic anion exchanger: Quaternary ammonium (Q)
Weak basic anion exchanger: Diethylamine (D)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty