Would you like to log in again?
Vi-CELL BLU Reagent Kits, Beckman Coulter
Supplier: Beckman Coulter
C39291 C06019 C06019 C39291
76409-827EA 510 USD InStock
76409-827 76409-829 CA76409-827 CA76409-829
Vi-CELL BLU Reagent Kits, Beckman Coulter
Assays Cellular Assays
We have improved the Vi-CELL reagent pack to make it easier to use. The Vi-CELL reagents you know and trust are now packaged in a convenient ready-to-load kit. Simply open the instrument reagent panel and slide in the reagent pack. Each pack can be used for 310 activities before changing. Packs are easy to replace with no mess. Say goodbye to wrangling with tubes and splattering dye when you change out reagents.
Each pack also includes an RFID tag that tells you how many activities are remaining on the pack. No more guessing on remaining reagent amounts or running out in the middle of a critical series of tests.
The embedded RFID tag gives important information about each pack to your Vi-CELL BLU instrument: Lot number, expiration date, on-board life (90 days), activities remaining and part number.
Each reagent pack contains: Trypan blue bottle, waste bottle, buffer solution bottle, disinfectant bottle and cleaning agent bottle.
Ordering information: C06019 is for a single Vi-CELL BLU Reagent pack. C39291 is for a quad pack (Quantity 4) of Vi-CELL BLU reagent packs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty