Would you like to log in again?
SHELF REINFORCED F/GROWTH CHAMBER KBW240
Supplier: BINDER INC
Shelf, reinforced - positioned at bottom level, max. Load 67 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system, for Growth chambers with light, Model KBW 240
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHELF REINFORCED F/GROWTH CHAMBER KBW240 76410-022 8012-1429 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty