Would you like to log in again?
ALARM OUTPUT ZERO VOLTAGE F/KBW 240/400
Supplier: BINDER INC
Alarm output, zero-voltage - for temperature (+/-2 degree C) and humidity (+/-5 % RH), accessible via 6-pin DIN socket (max. 24 V - 2.5 A), with audible signal that can be switched off, for model KBW 240/400(Growth chamber with light)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ALARM OUTPUT ZERO VOLTAGE F/KBW 240/400 76410-054 8012-1757 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty