Would you like to log in again?
PT 100 TEMPERATURE SENSOR FOR KBWF720
Supplier: BINDER INC
Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display, for Model KBWF 720(Growth chambers with light and humidity)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PT 100 TEMPERATURE SENSOR FOR KBWF720 76410-124 8012-1749 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty