Would you like to log in again?
VIEWING WINDOW/INTERIOR LIGHTING F/FD 56
Supplier: BINDER INC
Viewing window and interior lighting - Voltage: 120 V option model, viewing window (triple insulating glass) in door, 240 x 240 mm, and 15 W interior, for model fd 56(Drying and heating chamber)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIEWING WINDOW/INTERIOR LIGHTING F/FD 56 76410-344 8012-1702 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty