Would you like to log in again?
INERT GAS CONNECTION 10 MM FOR FD260
Supplier: BINDER INC
Inert gas connection - With gas inlet and outlet, incl. Additional measures for the greatest possible impermeability to gas, not in conjunction with access port or interior lighting option, for model fd260( drying and heating chamber), Diameter: 10 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INERT GAS CONNECTION 10 MM FOR FD260 76410-380 8012-1693 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty