Would you like to log in again?
EDC Pocket Knife, Slice®
Supplier: Slice
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
10496
76411-196CS 180.52 USD InStock
76411-196
EDC Pocket Knife, Slice®
Knives
This sleek, portable tool brings safety to the Everyday Carry community. It features the Slice® patent-pending grinding process, which creates a finger-friendly® edge that is safe to the touch and effective at cutting a wide variety of materials. The handle has a textured surface for a comfortable grip.
  • Manual retraction
  • Ambidextrous design
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Oil- and lubricant-free blade
  • Blade edge lasts up to 11.2× longer than metal
  • Vinyl-dipped hex key included
This sleek, portable tool brings safety to the EDC (Everyday Carry) community. It features the Slice® patent-pending grinding process, which creates a finger-friendly® edge that is both safe to the touch and effective at cutting a wide variety of materials. The handle is made from durable die-cast aluminum and offers a textured surface for a comfortable grip. An integrated lanyard hole and pocketable form factor keep the 10496 conveniently close and ready to go. It ships with a rounded-tip 10524 blade and is compatible with the pointed-tip 10525 and the serrated 10523. All compatible blades are made from advanced ceramics and last up to 11 times longer than equivalent metal blades. Custom blade-change hex key included.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty