Would you like to log in again?
Long-Handled Scraper With Safety Blade, Slice®
Supplier: Slice
10599
76411-198CS 308.96 USD InStock
76411-198
Long-Handled Scraper With Safety Blade, Slice®
Scrapers
The Slice® Long-Handled Scraper extends your reach with a sturdy aluminum handle. This scraper has a weight that’s balanced to feel solid without being too heavy to use comfortably.
  • Durable metal handle with comfort grip
  • Ambidextrous design
  • Finger-friendly® blade scrapes effectively
  • Reduces injuries; lowers costs
  • Chemically inert blades never rusts
  • Oil- and lubricant-free blade
  • Blade edge lasts up to 11.2× longer than metal
  • Cutting depth: 7.2 mm (0.28")
The angled blade encourages a natural wrist and arm position to improve ergonomics.
It uses Slice industrial blades. These 12 cm / 4" wide blades increase the scraping area, thus tackling larger jobs more efficiently. All Slice blades are made from non-rusting 100-percent zirconium oxide and last up to 11 times longer than comparable metal blades. They feature a finger-friendly® grinding process—which means the edge is much safer than traditional, dangerously sharp blades.
This scraper’s installed blade features rounded tips. It is also compatible with the 76314-920 Industrial Blades (Pointed Tip) and the 76411-178 Industrial Blades (Serrated). Slice’s largest scraper effectively removes paint, epoxies, and adhesives from smooth surfaces.
Accessories information: Includes 76314-918 blade. Slice®’s longest blade yet cuts through foam and fiberglass insulation like butter, without sacrificing user safety. With our patent-pending manufacturing process, this finger-friendly® blade is much safer to touch than traditional blades and cuts effectively when installed in industrial knives designed by Slice. As with all our blades, this 100-percent zirconium oxide safety blade is non-sparking, non-conductive, chemically inert, and never rusts. And, like all Slice blades, it lasts up to 11 times longer than comparable steel blades. This model features rounded tips for added safety. Comes in a pack of four blades. Reduce injuries and lower costs with Slice safety blades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty