Would you like to log in again?
VWR® Finger Textured Nitrile Examination Gloves
Supplier: VWR
VWR®
76411-510 76411-502 76411-504 76411-506 76411-508
76411-510CS 325.18 USD InStock
76411-510 76411-502 76411-508 76411-504 76411-506
VWR® Finger Textured Nitrile Examination Gloves
Gloves
Ambidextrous gloves are extra-soft and comfortable for everyday use. They protect hands from a broad range of chemicals.
  • Non sterile
  • Extra-soft and comfortable
  • Powder-free
  • Beaded cuff allows easy donning
Ideal for use in industrial and laboratory applications. Made of premium compounded nitrile for high manufacturing quality measures and product consistency. Gloves protect against biohazards and a broad range of chemicals. Composition is 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins. Finger textured gloves feature rolled, beaded cuffs.
Certifications: Gloves meet or exceed ASTM D6319 on water leak and dimension. Manufactured under GMP, ISO 9001, ISO 13485, and MDSAP quality system.
Packaging: 100 gloves per box, 10 boxes per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty