Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Puradisc Hydrophilic PTFE Syringe Filters, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Whatman™
6773-2504 6772-1302 6772-2504 6774-1302 6774-2502 6772-2502 6772-1302 6774-1304 6773-1302 6772-1304 6772-2502 6772-1304 6773-1302 6774-2504 6773-2504 6773-1304 6774-2504 6773-2502 6774-1302 6772-2504 6774-2502 6773-1304 6773-2502 6774-1304
76411-664PK 1408.85 USD InStock
76411-664 CA76411-652 76411-654 CA76411-662 76411-644 76411-666 CA76411-654 76411-656 CA76411-664 76411-660 76411-650 76411-662 CA76411-650 76411-652 CA76411-660 CA76411-648 76411-646 CA76411-656 76411-658 CA76411-644 CA76411-666 76411-648 CA76411-658 CA76411-646
Whatman™ Puradisc Hydrophilic PTFE Syringe Filters, Whatman products (Cytiva)
Filters Syringe Filters
Whatman Puradisc H-PTFE syringe filter contains a hydrophilic membrane that is chemically stable and inert. The H-PTFE membrane exhibits low protein binding and can be used for both aqueous and aggressive organic solvents.

  • Suitable for sample preparation for high performance and ultra-high performance liquid chromatography (HPLC and uHPLC)
  • Used with both aqueous solvents and aggressive organic solvents
  • Choice of 13 or 25 mm syringe filter diameter to suit sample volume
  • Choice of 0.2 or 0.45 μm pore sizes for retaining a variety of particles
  • Suitable for rapid syringe filtration of samples up to 100 ml
  • H-PTFE syringe filters combine high quality with cost-efficiency

Whatman Puradisc line is a comprehensive syringe filter portfolio used in a variety of applications, including capillary electrophoresis, biological preparations, protein filtration, and UV spectroscopy sample preparation.

Puradisc filters are available in a range of diameters and pore sizes, and eleven different material options to support diverse chemical solvents, compounds and sample types. Additional options include sterile, medical grade blister pack and tube tip format for accurate dispensing into a micro vial.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty