Would you like to log in again?
Nalgene® Biotainer Bottles, PC, Clean, Platinum Certified, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
CE-N3410-42 CE-N3405-42 CE-N3500-05 CE-N3500-20 CE-N3233-42 CE-N3120-42 CE-N3030-42
76412-176CS 1299.29 USD InStock
76412-176 76397-872 76397-874 76397-910 76397-876 76397-912 76397-878
Nalgene® Biotainer Bottles, PC, Clean, Platinum Certified, Thermo Scientific
Bottles Plastic Bottles
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Durable, translucent PC construction ensures the integrity of samples
  • Closures feature a silicone liner for leakproof assurance
  • Customization options are available for fluid transfer applications
  • Extensive validation documentation support
  • Suitable for use in storage conditions of –100 to 100 °C
Bottles are washed with USP water for injection and handled post-washing in Class 5 cleanrooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than ¹/₃ the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10⁻⁶ inside the container/closure, including two heat-sealed sterile barriers to protect the product prior to use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty