Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Secura® Balances, Standard Models, 100 - 240 V, Sartorius
Supplier: Sartorius
Secura®
SECURA213-1S SECURA6102-1S SECURA612-1S SECURA313-1S SECURA612-1S SECURA3102-1S SECURA613-1S SECURA225D-1S SECURA5102-1S SECURA224-1S SECURA1103-1S SECURA2102-1S SECURA613-1S SECURA124-1S SECURA2102-1S SECURA3101-1S SECURA124-1S SECURA324-1S SECURA1102-1S SECURA1102-1S SECURA125-1S SECURA213-1S SECURA3102-1S SECURA6101-1S SECURA6101-1S SECURA125-1S SECURA224-1S SECURA3101-1S SECURA6102-1S SECURA513-1S SECURA313-1S SECURA1103-1S SECURA5102-1S SECURA225D-1S SECURA513-1S SECURA324-1S
76412-438EA 5208.44 USD InStock
CA101485-972 CA101485-974 CA101485-976 76412-438 CA101485-978 76412-436 76412-458 76412-442 76412-464 76412-440 76412-462 76412-468 76412-446 76412-444 CA76412-452 76412-466 CA102355-796 CA490019-380 CA102355-798 76412-460 CA101485-980 CA490019-378 CA101485-982 CA101485-984 CA101485-986 CA76412-436 CA76412-458 CA101486-028 76412-448 76412-452 76412-456 76412-434 76412-454 CA76412-464 CA102355-764 76412-470
Secura® Balances, Standard Models, 100 - 240 V, Sartorius
Balances and Scales
Secura is a laboratory balance that does far more than set new standards in metrological specifications.
  • LevelControl Keeps Balance Correctly Leveled
  • No Risk Due to Temperature Fluctuations
  • Stress-Free USP Compliant Measurements
  • Password Protection
  • Interactive User Guidance for Fast GLP and GMP-Compliant Printouts
It also features built-in protection systems for complete reliability and regulatory compliance.
Real-time guidance prompts for leveling, automatic internal adjustment, and 100% traceable, clear documentation with sample and batch identifiers make lab work more efficient.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty