Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
WEIGHT SET 200G 20G 20MG ASTM CLASS 1
Supplier: SARTORIUS CORPORATION EDI
Cal-Pak* Weight set, ASTM Class 1 with NVLAP Certificate, Contains three high quality precision weights 200G 20G 20MG, Can be used with all balance, Supplied in a rugged polycarbonate case with each individual weight in its own high quality polycarbonate case
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WEIGHT SET 200G 20G 20MG ASTM CLASS 1 76412-514 WCP281AC1C Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty