Would you like to log in again?
CALIBRATION WEIGHT CLASS 1 W/NVLAP 100MG
Supplier: SARTORIUS CORPORATION EDI
Calibration Weight, Individual, with Nvlap certificate(National Voluntary Laboratory Accreditation Program)evaluates and accredits laboratories in their respective fields and ensures they meet ISO standard, ANSI/ASTM E617 -Class 1, Certificates determine the price of calibration weight, size: 100mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CALIBRATION WEIGHT CLASS 1 W/NVLAP 100MG 76412-726 WI100MAC1C Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty