Would you like to log in again?
Data Printer for Sartorius Balances, Sartorius
Supplier: SARTORIUS CORPORATION EDI
YDP20-0CE YDP20-0CE
76412-970EA 1574.46 USD InStock
76412-970 CA97021-988
Data Printer for Sartorius Balances, Sartorius
Printers Document Printers
This data printer suitable for external verification with date, time, statistics, numerator.
  • Values transferred from an on-line weighing instrument are stored
In addition to recording individual weighing data, the data printer can generate a statistical evaluation of weighing data.
Certifications: This data printer meets the quality standards of the chemical and pharmaceutical industries, in compliance with the requirements of GLP (Good Laboratory Practice) and GMP (Good Manufacturing Practice).
Delivery information: Includes cable.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty