Would you like to log in again?
SAVE-LOCK TUBE HOLDER 2ML MICRO-BALANCES
Supplier: SARTORIUS CORPORATION EDI
Save-Lock Tube Holder, for micro-balances, enables easy and safe positioning of Eppendorf tubes and centrifuge reaction tubes on the balance, Volume: up to 2ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAVE-LOCK TUBE HOLDER 2ML MICRO-BALANCES 76413-028 YSH13 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty