Would you like to log in again?
Plant Growth Chambers, KBW Series, Binder
Supplier: Binder
9020-0340 9020-0338 9020-0339
76413-530EA 19274.8 USD InStock
76413-530 76413-532 76413-534
Plant Growth Chambers, KBW Series, Binder
Testing Chambers Plant Growth Chambers
The KBW series plant growth chambers are precise units for plant cultivation under advanced working conditions. They provide perfect simulation of a wide range of growing environments. The units satisfy all requirements for optimum lighting and temperature conditions, so that culture processes can be defined and reproduced exactly. The design includes two or three variable position daylight cassettes with five daylight luminescence tubes each, which provides homogeneous light distribution. Units self-test for comprehensive status analysis.
  • Precision and outstanding dynamics
  • Even distribution of light on all levels and adjustable fan speed
  • Environmentally friendly
  • Intuitive touchscreen controller with time-segment and real-time programming
  • Ethernet interface for APT-COM™ DataControlSystem communication software
The KBW models have a heated inner glass door, an access port with silicone plug Ø30 mm on the left hand side. Units feature an independent adjustable temperature safety device Class 3.1 with visual and audible alarm, which protects the samples and the incubator. Microprocessor controller with two programs each with 10 sections or, alternatively, switch over to one program with 20 sections. Digital temperature setting with an accuracy of a tenth of a degree, integrated weekly program timer, elapsed time indicator and electronically controlled APT.line™ preheating chamber ensures temperature accuracy and reproducible results.
Values measured via the internal data logger can be read out in open format via USB. All models have an tight sealing inner door made of ESG safety glass, the door is heated against condensation. Glass corrosion is avoided thanks to the special TIMELESS coating. The units have 4 casters each, 2 with brakes them.
Temperature fluctuation (time) (°C): ±0.2 (without illumination), ±0.1 (with 100% illumination)
Temperature variation (spatial) (°C): ±0.5 (without illumination), ±0.5 (with 100% illumination)
Delivery information: Supplied with two (or three) stainless steel shelves and Binder test certificate. Calibration and validation is possible, for details please contact your local Avantor sales office.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty